2,3-Difluoro-4-(difluoromethyl)pyridine 95%

Leverandør: Apollo Scientific
Warning

PC49147-10G PC49147-1G PC49147-5G
APOSPC49147-10GEA 22850 NOK
APOSPC49147-10G APOSPC49147-1G APOSPC49147-5G
2,3-Difluoro-4-(difluoromethyl)pyridine 95%
2,3-Difluoro-4-(difluoromethyl)pyridine
Formel: C₆H₃F₄N
MW: 165.09 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD22199287
CAS-nummer: 1374659-38-7
FN: 1993
ADR: 3 III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR