1-Benzyloxy-4-(difluoromethyl)benzene 98%

Leverandør: Apollo Scientific
Warning

Synonymer: 1-(Benzyloxy)-4-(difluoromethyl)benzene

PC49551-1G
APOSPC49551-1GEA 1275 NOK
APOSPC49551-1G
1-Benzyloxy-4-(difluoromethyl)benzene 98%
1-Benzyloxy-4-(difluoromethyl)benzene
Formel: C₁₄H₁₂F₂O
MW: 234,25 g/mol
Smeltepunkt: 74…76 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD18803519
CAS-nummer: 915799-67-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR