2-(Difluoromethoxy)-5-fluorobenzeneboronic acid, pinacol ester 95%

Leverandør: Apollo Scientific
Warning

Synonymer: 2-(2-(Difluoromethoxy)-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PC49715-1G PC49715-250MG PC49715-5G
APOSPC49715-1GEA 12100 NOK
APOSPC49715-1G APOSPC49715-250MG APOSPC49715-5G
2-(Difluoromethoxy)-5-fluorobenzeneboronic acid, pinacol ester 95%
2-(Difluoromethoxy)-5-fluorobenzeneboronic acid, pinacol ester
Formel: C₁₃H₁₆BF₃O₃
MW: 288.071 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD30486443
CAS-nummer: 1417301-27-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR