Litiumfluorid 99.99%

Leverandør: Apollo Scientific
Danger

PC4990R-100G
APOSPC4990R-100GEA 7975 NOK
APOSPC4990R-100G
Litiumfluorid 99.99%
Litiumfluorid
Formel: FLi
Kokepunkt: 1680 °C
Smeltepunkt: 845 °C
Tetthet: 2,635 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011090
CAS-nummer: 7789-24-4
EINECS: 232-152-0
FN: 3288
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR