3-Fluoro-4-(hydroxymethyl)benzonitrile 99+%

Leverandør: Apollo Scientific
Danger

Synonymer: 4-Cyano-2-fluorobenzyl alcohol

PC500665-1G PC500665-5G PC500665-250MG
APOSPC500665-1GEA 305 NOK
APOSPC500665-1G APOSPC500665-5G APOSPC500665-250MG
3-Fluoro-4-(hydroxymethyl)benzonitrile 99+%
3-Fluoro-4-(hydroxymethyl)benzonitrile
Formel: C₈H₆FNO
MW: 151.138 g/mol
Smeltepunkt: 63…68 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09037779
CAS-nummer: 219873-06-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR