2,6-Difluoro-m-toluidine 98%

Leverandør: Apollo Scientific
Warning

PC51074-10G PC51074-5G PC51074-1G
APOSPC51074-10GEA 3775 NOK
APOSPC51074-10G APOSPC51074-5G APOSPC51074-1G
2,6-Difluoro-m-toluidine 98%
2,6-Difluoro-m-toluidine
Formel: C₇H₇F₂N
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD06660182
CAS-nummer: 144851-63-8
FN: 2810
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR