Ethyl 5-amino-4-bromo-1-[4-(trifluoromethyl)phenyl]pyrazole-3-carboxylate

Leverandør: Apollo Scientific

PC520009-1G
APOSPC520009-1GEA 11225 NOK
APOSPC520009-1G
Ethyl 5-amino-4-bromo-1-[4-(trifluoromethyl)phenyl]pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-[4-(trifluoromethyl)phenyl]pyrazole-3-carboxylate
Formel: C₁₃H₁₁BrF₃N₃O₂
MW: 378,145 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD23135140
CAS-nummer: 1427022-87-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR