Ethyl 5-amino-1-[2-(trifluoromethyl)phenyl]pyrazole-3-carboxylate

Leverandør: Apollo Scientific

PC520011-1G
APOSPC520011-1GEA 11775 NOK
APOSPC520011-1G
Ethyl 5-amino-1-[2-(trifluoromethyl)phenyl]pyrazole-3-carboxylate
Ethyl 5-amino-1-[2-(trifluoromethyl)phenyl]pyrazole-3-carboxylate
Formel: C₁₃H₁₂F₃N₃O₂
MW: 299,248 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD07388456
CAS-nummer: 866838-05-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR