2-Chloro-1-[5-(difluoromethyl)-1H-pyrrol-3-yl]ethanone

Leverandør: Apollo Scientific

PC520174-1G
APOSPC520174-1GEA 33700 NOK
APOSPC520174-1G
2-Chloro-1-[5-(difluoromethyl)-1H-pyrrol-3-yl]ethanone
2-Chloro-1-[5-(difluoromethyl)-1H-pyrrol-3-yl]ethanone
Formel: C₇H₆ClF₂NO
MW: 193,578 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD29767654
CAS-nummer: 2091118-89-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR