1-(Difluoromethyl)-3-methyl-benzene

Leverandør: Apollo Scientific

PC520310-1G
APOSPC520310-1GEA 7725 NOK
APOSPC520310-1G
1-(Difluoromethyl)-3-methyl-benzene
1-(Difluoromethyl)-3-methyl-benzene
Formel: C₈H₈F₂
MW: 142,146 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD29767779
CAS-nummer: 1637371-42-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR