Methyl 5-bromo-2-(difluoromethyl)benzoate

Leverandør: Apollo Scientific

PC520315-1G
APOSPC520315-1GEA 27850 NOK
APOSPC520315-1G
Methyl 5-bromo-2-(difluoromethyl)benzoate
Methyl 5-bromo-2-(difluoromethyl)benzoate
Formel: C₉H₇BrF₂O₂
MW: 265,051 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD30496628
CAS-nummer: 1630983-08-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR