1-(2-Fluoro-4-pyridyl)cyclopropanecarboxylic acid

Leverandør: Apollo Scientific

PC520384-1G
APOSPC520384-1GEA 33700 NOK
APOSPC520384-1G
1-(2-Fluoro-4-pyridyl)cyclopropanecarboxylic acid
1-(2-Fluoro-4-pyridyl)cyclopropanecarboxylic acid
Formel: C₉H₈FNO₂
MW: 181,164 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD23134927
CAS-nummer: 1427021-97-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR