1-(Difluoromethoxy)-3-(trifluoromethyl)benzene 95%

Leverandør: Apollo Scientific

PC520707-10G
APOSPC520707-10GEA 28425 NOK
APOSPC520707-10G
1-(Difluoromethoxy)-3-(trifluoromethyl)benzene 95%
1-(Difluoromethoxy)-3-(trifluoromethyl)benzene
Formel: C₈H₅F₅O
MW: 212,117 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD14698439
CAS-nummer: 119558-79-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR