4-(4-Fluorophenoxy)piperidine hydrochloride 97%

Leverandør: Apollo Scientific

PC530028-1G PC530028-5G
APOSPC530028-1GEA 3850 NOK
APOSPC530028-1G APOSPC530028-5G
4-(4-Fluorophenoxy)piperidine hydrochloride 97%
4-(4-Fluorophenoxy)piperidine hydrochloride
Formel: C₁₁H₁₅ClFNO
MW: 231.690 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD04115015
CAS-nummer: 3202-34-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR