4,5-Difluoro-o-toluidine 95%

Leverandør: Apollo Scientific
Warning

PC53402-25G PC53402-5G PC53402-1G
APOSPC53402-25GEA 4575 NOK
APOSPC53402-25G APOSPC53402-5G APOSPC53402-1G
4,5-Difluoro-o-toluidine 95%
4,5-Difluoro-o-toluidine
Formel: C₇H₇F₂N
Smeltepunkt: 56…58 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD08062378
CAS-nummer: 875664-57-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR