(S)-6-Fluorochromane-2-carboxylic acid ≥97%

Leverandør: Apollo Scientific

Synonymer: (S)-6-Fluorochroman-2-carboxylic acid

PC540014-1G
APOSPC540014-1GEA 1375 NOK
APOSPC540014-1G
(S)-6-Fluorochromane-2-carboxylic acid ≥97%
(S)-6-Fluorochromane-2-carboxylic acid
Formel: C₁₀H₉FO₃
MW: 196,175 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD20487794
CAS-nummer: 129101-36-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR