5-Bromo-2-(difluoromethoxy)-4-methylpyridine ≥98%

Leverandør: Apollo Scientific

PC540038-250MG
APOSPC540038-250MGEA 1400 NOK
APOSPC540038-250MG
5-Bromo-2-(difluoromethoxy)-4-methylpyridine ≥98%
5-Bromo-2-(difluoromethoxy)-4-methylpyridine
Formel: C₇H₆BrF₂NO
MW: 238.029 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD14698387
CAS-nummer: 1079352-13-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR