3-(Dimethoxymethyl)-4-[3-(trifluoromethyl)phenoxy]aniline 95%

Leverandør: Apollo Scientific

PC56289-5G
APOSPC56289-5GEA 14425 NOK
APOSPC56289-5G
3-(Dimethoxymethyl)-4-[3-(trifluoromethyl)phenoxy]aniline 95%
3-(Dimethoxymethyl)-4-[3-(trifluoromethyl)phenoxy]aniline
Formel: C₁₆H₁₆F₃NO₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD31614284
CAS-nummer: 2244088-17-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR