4-Fluorophenylglyoxal hydrate

Leverandør: Apollo Scientific
Warning

Synonymer: 2-(4-Fluorophenyl)-2-oxoacetaldehyde hydrate, 4-Fluorophenyl(oxo)glyoxal hydrate

PC5633-1G PC5633-5G PC5633-10G
APOSPC5633-1GEA 270 NOK
APOSPC5633-1G APOSPC5633-5G APOSPC5633-10G
4-Fluorophenylglyoxal hydrate
4-Fluorophenylglyoxal hydrate
Formel: C₈H₇FO₃
Kokepunkt: 115 °C/35mmHg
Smeltepunkt: 80…88 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01320770
CAS-nummer: 447-43-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR