3-((Trifluoromethyl)thio)benzoic acid 98%

Leverandør: Apollo Scientific
Warning

PC5639-1G PC5639-25G PC5639-5G
APOSPC5639-1GEA 450 NOK
APOSPC5639-1G APOSPC5639-25G APOSPC5639-5G
3-((Trifluoromethyl)thio)benzoic acid 98%
3-((Trifluoromethyl)thio)benzoic acid
Formel: C₈H₅F₃O₂S
Smeltepunkt: 71…74 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00236337
CAS-nummer: 946-65-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR