4-Fluorophenylboronic acid pinacol ester 97%

Leverandør: Apollo Scientific

Synonymer: 2-(4-Fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Fluorophenylboronic acid Pinacol ester, 1-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

PC5659-100G
APOSPC5659-100GEA 1100 NOK
APOSPC5659-100G
4-Fluorophenylboronic acid pinacol ester 97%
4-Fluorophenylboronic acid pinacol ester
Formel: C₁₂H₁₆BFO₂
MW: 222.0637 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03453665
CAS-nummer: 214360-58-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR