2-Fluoro-5-(hydroxymethyl)benzonitrile

Leverandør: Apollo Scientific
Warning

PC6132-1G PC6132-5G PC6132-25G
APOSPC6132-1GEA 270 NOK
APOSPC6132-1G APOSPC6132-5G APOSPC6132-25G
2-Fluoro-5-(hydroxymethyl)benzonitrile
2-Fluoro-5-(hydroxymethyl)benzonitrile
Formel: C₈H₆FNO
MW: 151.14 g/mol
Smeltepunkt: 45…47 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03094340
CAS-nummer: 227609-85-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR