5-(Cyanomethyl)-2-fluorobenzonitrile 95+%

Leverandør: Apollo Scientific

PC6135-1G
APOSPC6135-1GEA 985 NOK
APOSPC6135-1G
5-(Cyanomethyl)-2-fluorobenzonitrile 95+%
5-(Cyanomethyl)-2-fluorobenzonitrile
Formel: C₉H₅FN₂
MW: 160.1500 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03094342
CAS-nummer: 519059-09-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR