4-((Trifluoromethyl)thio)benzoic acid 98%

Leverandør: Apollo Scientific
Warning

PC6204-100G PC6204-25G PC6204-5G
APOSPC6204-100GEA 1825 NOK
APOSPC6204-100G APOSPC6204-25G APOSPC6204-5G
4-((Trifluoromethyl)thio)benzoic acid 98%
4-((Trifluoromethyl)thio)benzoic acid
Formel: C₈H₅F₃O₂S
Smeltepunkt: 159…163 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00040906
CAS-nummer: 330-17-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR