α-Bromo-3-(difluoromethoxy)toluene 98%

Leverandør: Apollo Scientific
Danger

Synonymer: 3-(Difluoromethoxy)benzyl bromide, 1-(Bromomethyl)-3-(difluoromethoxy)benzene, m-(Difluoromethoxy)benzyl bromide

PC6823-500MG PC6823-1G PC6823-5G
APOSPC6823-500MGEA 270 NOK
APOSPC6823-500MG APOSPC6823-1G APOSPC6823-5G
α-Bromo-3-(difluoromethoxy)toluene 98%
α-Bromo-3-(difluoromethoxy)toluene
Formel: C₈H₇BrF₂O
MW: 237.04 g/mol
Kokepunkt: 76 °C/1mmHg
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00236232
CAS-nummer: 72768-95-7
FN: 3265
ADR: 8 II
New Product

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR