5-Bromo-2-fluorobenzeneboronic acid pinacol ester

Leverandør: Apollo Scientific

Synonymer: 5-Bromo-2-fluorophenylboronic acid pinacol ester

PC6981-5G PC6981-25G
APOSPC6981-5GEA 1625 NOK
APOSPC6981-5G APOSPC6981-25G
5-Bromo-2-fluorobenzeneboronic acid pinacol ester
5-Bromo-2-fluorobenzeneboronic acid pinacol ester
Formel: C₁₂H₁₅BBrFO₂
MW: 300.96 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09260441
CAS-nummer: 942069-51-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR