2-Cyano-4-fluorophenylboronic acid pinacol ester

Leverandør: Apollo Scientific
Warning

Synonymer: 2-Cyano-4-fluorophenylboronic Acid Pinacol Ester, 2-Cyano-4-fluorobenzeneboronic acid pinacol ester

PC7020-5G
APOSPC7020-5GEA 2075 NOK
APOSPC7020-5G
2-Cyano-4-fluorophenylboronic acid pinacol ester
2-Cyano-4-fluorophenylboronic acid pinacol ester
Formel: C₁₃H₁₅BFNO₂
MW: 247.07 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09260444
CAS-nummer: 461451-63-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR