4-Bromomethyl-3-fluorobenzeneboronic acid pinacol ester 95+%

Leverandør: Apollo Scientific
Warning

Synonymer: 4-Bromomethyl-3-fluorophenylboronic acid pinacol ester, 2-(4-(Bromomethyl)-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-Dioxaborolane, 2-[4-(bromomethyl)-3-fluorophenyl]-4,4,5,5-tetramethyl-, 2-[4-(Bromomethyl)-3-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PC7518-5G
APOSPC7518-5GEA 3350 NOK
APOSPC7518-5G
4-Bromomethyl-3-fluorobenzeneboronic acid pinacol ester 95+%
4-Bromomethyl-3-fluorobenzeneboronic acid pinacol ester
Formel: C₁₃H₁₇BBrFO₂
MW: 314.99 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD11855987
CAS-nummer: 1150271-74-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR