α,α,α-Trifluoro-p-toluic acid 98%

Leverandør: Apollo Scientific
Warning

Synonymer: 4-(Trifluoromethyl)benzoic acid

PC7540-100G PC7540-25G PC7540-500G
APOSPC7540-100GEA 305 NOK
APOSPC7540-100G APOSPC7540-25G APOSPC7540-500G
α,α,α-Trifluoro-p-toluic acid 98%
α,α,α-Trifluoro-p-toluic acid
Formel: C₈H₅F₃O₂
MW: 190.12 g/mol
Kokepunkt: 247 °C/753mmHg
Smeltepunkt: 219…222 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002562
CAS-nummer: 455-24-3
EINECS: 207-242-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR