1-[2,6-Bis(trifluoromethyl)quinol-4-yl]piperazine 97%

Leverandør: Apollo Scientific
Warning

PC7615-500MG
APOSPC7615-500MGEA 1175 NOK
APOSPC7615-500MG
1-[2,6-Bis(trifluoromethyl)quinol-4-yl]piperazine 97%
1-[2,6-Bis(trifluoromethyl)quinol-4-yl]piperazine
Formel: C₁₅H₁₃F₆N₃
MW: 349,274 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03094433
CAS-nummer: 401567-74-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR