2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine

Leverandør: Apollo Scientific
Warning

Synonymer: 2,4,6-Tris(perfluoropropyl)-1,3,5-triazine

PC7890-25G
APOSPC7890-25GEA 11200 NOK
APOSPC7890-25G
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine

Mass spectrometry standard.

Formel: C₁₂F₂₁N₃
MW: 585.12 g/mol
Kokepunkt: 165 °C
Tetthet: 1.716 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00042438
CAS-nummer: 915-76-4

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR