Ultramark®

Leverandør: Apollo Scientific
Warning

Synonymer: SpectraSynQ PFPE-1600

PC7955-1G
APOSPC7955-1GEA 1550 NOK
APOSPC7955-1G
Ultramark®
Ultramark®

Fluorinated polyether for Mass Spectrometry up tomass 3200

Kokepunkt: >210 °C/0.7mmHg
Tetthet: 1.8 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00166131
CAS-nummer: 66795-94-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR