4-(Trifluoromethylthio)phenylacetic acid 96%

Leverandør: Apollo Scientific
Warning

PC8253-25G PC8253-5G PC8253-1G
APOSPC8253-25GEA 3775 NOK
APOSPC8253-25G APOSPC8253-5G APOSPC8253-1G
4-(Trifluoromethylthio)phenylacetic acid 96%
4-(Trifluoromethylthio)phenylacetic acid
Formel: C₉H₇F₃O₂S
Smeltepunkt: 114…118 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00236351
CAS-nummer: 243977-23-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR