3-Fluoro-4-propoxybenzeneboronic acid

Leverandør: Apollo Scientific
Warning

Synonymer: (3-Fluoro-4-propoxyphenyl)boronic acid, 3-Fluoro-4-propoxyphenylboronic acid

PC8650-1G PC8650-5G PC8650-25G
APOSPC8650-1GEA 270 NOK
APOSPC8650-1G APOSPC8650-5G APOSPC8650-25G
3-Fluoro-4-propoxybenzeneboronic acid
3-Fluoro-4-propoxybenzeneboronic acid
Formel: C₉H₁₂BFO₃
MW: 197.999 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD06411285
CAS-nummer: 192376-68-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR