3-Fluoroisonicotinic acid

Leverandør: Apollo Scientific
Warning

Synonymer: 3-Fluoropyridine-4-carboxylic acid

PC8767-25G
APOSPC8767-25GEA 790 NOK
APOSPC8767-25G
3-Fluoroisonicotinic acid
3-Fluoroisonicotinic acid
Formel: C₆H₄FNO₂
MW: 141.1 g/mol
Smeltepunkt: 251…254 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03789251
CAS-nummer: 393-53-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR