2-(Aminomethyl)-6-fluoropyridine dihydrochloride

Leverandør: Apollo Scientific
Danger

PC8931-100MG
APOSPC8931-100MGEA 270 NOK
APOSPC8931-100MG
2-(Aminomethyl)-6-fluoropyridine dihydrochloride
2-(Aminomethyl)-6-fluoropyridine dihydrochloride
Formel: C₆H₇FN₂·2ClH
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD16872044
CAS-nummer: 1257535-08-2
FN: 3261
ADR: 8,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR