2-(Cyclopropylmethoxy)-5-fluorobenzonitrile 95%

Leverandør: Apollo Scientific

PC900219-1G
APOSPC900219-1GEA 4475 NOK
APOSPC900219-1G
2-(Cyclopropylmethoxy)-5-fluorobenzonitrile 95%
2-(Cyclopropylmethoxy)-5-fluorobenzonitrile
Formel: C₁₁H₁₀FNO
MW: 191.202 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD18860667
CAS-nummer: 1342025-16-4

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR