3-Fluoro-4-pyrrolidin-1-ylbenzaldehyde 96%

Leverandør: Apollo Scientific

PC901565-1G
APOSPC901565-1GEA 4050 NOK
APOSPC901565-1G
3-Fluoro-4-pyrrolidin-1-ylbenzaldehyde 96%
3-Fluoro-4-pyrrolidin-1-ylbenzaldehyde
Formel: C₁₁H₁₂FNO
MW: 193,22 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD05237677
CAS-nummer: 153659-43-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR