[1-(4-Fluorophenyl)-1H-pyrazol-4-yl]methanol 97%

Leverandør: Apollo Scientific

PC902881-1G
APOSPC902881-1GEA 6425 NOK
APOSPC902881-1G
[1-(4-Fluorophenyl)-1H-pyrazol-4-yl]methanol 97%
[1-(4-Fluorophenyl)-1H-pyrazol-4-yl]methanol
Formel: C₁₀H₉FN₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD08059807
CAS-nummer: 153863-34-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR