2-Fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzonitrile 97%

Leverandør: Apollo Scientific

PC905878-5G
APOSPC905878-5GEA 1050 NOK
APOSPC905878-5G
2-Fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzonitrile 97%
2-Fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzonitrile
Formel: C₁₆H₁₀FN₃O
MW: 279.269 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD28133450
CAS-nummer: 1021298-68-9

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR