4-Acetyl-2-fluorobenzonitrile 97%

Leverandør: Apollo Scientific

PC907486-1G
APOSPC907486-1GEA 1200 NOK
APOSPC907486-1G
4-Acetyl-2-fluorobenzonitrile 97%
4-Acetyl-2-fluorobenzonitrile
Formel: C₉H₆FNO
MW: 163.148 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD10575008
CAS-nummer: 214760-18-6

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR