2,6-Difluorocinnamic aldehyde 95%

Leverandør: Apollo Scientific

PC908912-1G
APOSPC908912-1GEA 3200 NOK
APOSPC908912-1G
2,6-Difluorocinnamic aldehyde 95%
2,6-Difluorocinnamic aldehyde
Formel: C₉H₆F₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD07782064
CAS-nummer: 117338-43-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR