3-(2-Fluorophenyl)-5-methyl-1,2,4-oxadiazole 98%

Leverandør: Apollo Scientific

PC909131-1G
APOSPC909131-1GEA 3325 NOK
APOSPC909131-1G
3-(2-Fluorophenyl)-5-methyl-1,2,4-oxadiazole 98%
3-(2-Fluorophenyl)-5-methyl-1,2,4-oxadiazole
Formel: C₉H₇FN₂O
MW: 178,163 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD23699515
CAS-nummer: 1426958-50-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR