4-Chloro-2-fluoro-3-methylphenol 96%

Leverandør: Apollo Scientific

PC910237-1G
APOSPC910237-1GEA 7775 NOK
APOSPC910237-1G
4-Chloro-2-fluoro-3-methylphenol 96%
4-Chloro-2-fluoro-3-methylphenol
Formel: C₇H₆ClFO
MW: 160,573 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD20526390
CAS-nummer: 1351668-24-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR