4-Fluoro-3-(hydroxymethyl)benzonitrile 98%

Leverandør: Apollo Scientific

PC910950-1G PC910950-250MG
APOSPC910950-1GEA 7325 NOK
APOSPC910950-1G APOSPC910950-250MG
4-Fluoro-3-(hydroxymethyl)benzonitrile 98%
4-Fluoro-3-(hydroxymethyl)benzonitrile
Formel: C₈H₆FNO
MW: 151.138 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD11213689
CAS-nummer: 856931-47-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR