1,1,2,3,3,3-Hexafluoropropyl chlorofluoromethyl ether 97%

Leverandør: Apollo Scientific

PC9259-5G
APOSPC9259-5GEA 2600 NOK
APOSPC9259-5G
1,1,2,3,3,3-Hexafluoropropyl chlorofluoromethyl ether 97%
1,1,2,3,3,3-Hexafluoropropyl chlorofluoromethyl ether
Formel: C₄H₂ClF₇O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03094548
CAS-nummer: 56860-86-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR