2-Bromo-6-iodo-4-(trifluoromethyl)aniline 95+%

Leverandør: Apollo Scientific

Synonymer: 4-Amino-3-bromo-5-iodobenzotrifluoride

PC9570-1G
APOSPC9570-1GEA 750 NOK
APOSPC9570-1G
2-Bromo-6-iodo-4-(trifluoromethyl)aniline 95+%
2-Bromo-6-iodo-4-(trifluoromethyl)aniline
Formel: C₇H₄BrF₃IN
MW: 365,9170 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD08458149
CAS-nummer: 875306-20-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR