Lloyd and McCown's woody plant medium with vitamins
Leverandør: Apollo Scientific
PML449B-10L
APOSPML449B-10LEA 840 NOK
APOSPML449B-10L
Lloyd and McCown's woody plant medium with vitamins
Media, Cellekultur
Contains the WPM macro- and micronutrients and vitamins as described by Lloyd and McCown (1981). Plant tissue culture tested.