Vann BP, Ph. Eur., pharma grade

Leverandør: PanReac AppliChem

Synonymer: Hydrogen oxide

141074.1315
APPC141074.1315EA 1775 NOK
APPC141074.1315
Vann BP, Ph. Eur., pharma grade
Vann

Pharma grade water is ideal for laboratory utilisation, analysis, research and fine chemistry.

Formel: H₂O
MDL nummer: MFCD00011332
CAS-nummer: 7732-18-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR