EDTA dinatrium salt dihydrat 99.0-101.0% (by complexometric analysis) USP, BP, Ph. Eur., pharma grade

Leverandør: PanReac AppliChem
Warning

141669.0416 141669.0914 141669.1211
APPC141669.0416EA 31325 NOK
APPC141669.0416 APPC141669.0914 APPC141669.1211
EDTA dinatrium salt dihydrat 99.0-101.0% (by complexometric analysis) USP, BP, Ph. Eur., pharma grade
EDTA dinatrium salt dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Smeltepunkt: 248…254 °C
MDL nummer: MFCD00150037
CAS-nummer: 6381-92-6
EINECS: 205-358-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (Complexometric) 99.0 - 101.0%
Identity According to Pharmacopoeias Passes Test
pH of 5% Solution 4.0 - 5.0
Insoluble Matter in H₂O 0.01%
Loss on Drying at 150 °C (6 Hours) 8.7 - 11.4%
Chloride (Cl) 0.02%
Sulphate (SO₄) 0.1%
Residual Solvents (Ph. Eur./USP) Passes Test
Nitrilotriacetic Acid [(CH₂COOH)₃N] 0.1%
Heavy Metals (as Pb) 0.001%
Ca Passes Test
Fe 0.0005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR